Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,3,4-trisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,3,4-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD03412019
Synonym Ins(1,3,4)-P3; 1,3,4-IP3 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66963EXD

 
alfa aesar™
J66963EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66963EXZ

 
alfa aesar™
J66963EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66963MCR

 
alfa aesar™
J66963MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-1,3,4-trisphosphate sodium salt
MDL Number MFCD03412019
Physical Form Lyophilized Powder
Assay 98%
Synonym Ins(1,3,4)-P3; 1,3,4-IP3 sodium salt
Quantity 0.5mg
Color White to off-white
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C