missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt, 95%, Thermo Scientific Chemicals
$164.00 - $164.00
Chemical Identifiers
CAS | 142507-74-2 |
---|---|
Molecular Formula | C6H19NO18P4 |
Molecular Weight (g/mol) | 517.102 |
MDL Number | MFCD01459915 |
InChI Key | ANQVRWQIKKBLLK-ADHKWWJESA-N |
Synonym | myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt, 1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine |
PubChem CID | 71308569 |
IUPAC Name | azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate |
SMILES | C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66936EXZ
|
Thermo Scientific Chemicals
J66936EXZ |
0.1 mg |
Each for $164.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142507-74-2 | |
517.102 | |
ANQVRWQIKKBLLK-ADHKWWJESA-N | |
71308569 | |
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N |
C6H19NO18P4 | |
MFCD01459915 | |
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt, 1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine | |
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate |
Specifications
142507-74-2 | |
C6H19NO18P4 | |
0.1 mg | |
ANQVRWQIKKBLLK-ADHKWWJESA-N | |
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate | |
71308569 | |
95% | |
D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt |
White | |
MFCD01459915 | |
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt, 1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine | |
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N | |
517.102 | |
568.2 | |
Lyophilized Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only