missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66936EXZ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-myo-Inositol-1,3,4,6-tetrakis(phosphate) ammonium salt | |
White | |
MFCD01459915 | |
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt, 1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine | |
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N | |
517.102 | |
568.2 | |
Lyophilized Powder |
142507-74-2 | |
C6H19NO18P4 | |
0.1 mg | |
ANQVRWQIKKBLLK-ADHKWWJESA-N | |
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate | |
71308569 | |
95% |
Chemical Identifiers
142507-74-2 | |
517.102 | |
ANQVRWQIKKBLLK-ADHKWWJESA-N | |
71308569 | |
C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)O)OP(=O)(O)O)OP(=O)(O)O)O.N |
C6H19NO18P4 | |
MFCD01459915 | |
myo-inositol 1,3,4,6-tetrakis-phosphate ammonium salt, 1r,2s,3s,4s,5r,6r-2,5-dihydroxy-3,4,6-tris phosphonooxy cyclohexyl oxyphosphonic acid amine | |
azane;[(1S,3R,4R,6S)-2,5-dihydroxy-3,4,6-triphosphonooxycyclohexyl] dihydrogen phosphate |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only