Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
CAS 210488-61-2
Formula Weight 675.93
MDL Number MFCD00153818
Synonym Ins(1,3,4,5)P4 sodium salt; 1,3,4,5-IP4 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66461EXD

  SDS
alfa aesar™
J66461EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66461EXZ

  SDS
alfa aesar™
J66461EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 675.93
CAS 210488-61-2
MDL Number MFCD00153818
Assay 98%
Synonym Ins(1,3,4,5)P4 sodium salt; 1,3,4,5-IP4 sodium salt
Chemical Name or Material D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Physical Form Lyophilized Powder
Safety and Handling

-30° to -10°C