missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2-bisphosphate sodium salt, 98%

$180.00 - $810.03

Chemical Identifiers

CAS 208584-51-4
Molecular Formula C6H14Na2O12P2
Molecular Weight (g/mol) 386.093
MDL Number MFCD10567083
InChI Key PHZDHEPQFXTBRQ-JBGYRJLXSA-N
Synonym d-myo-inositol-1,2-diphosphate sodium salt
PubChem CID 131875843
IUPAC Name sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate
SMILES C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66420EXZ
View Documents
Alfa Aesar™
J66420EXZ
0.1mg
Each for $180.00
 
AAJ66420EXD
View Documents
Alfa Aesar™
J66420EXD
0.5mg
Each for $810.03
 
Description

Description

Specifications

Chemical Identifiers

208584-51-4
386.093
PHZDHEPQFXTBRQ-JBGYRJLXSA-N
131875843
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na]
C6H14Na2O12P2
MFCD10567083
d-myo-inositol-1,2-diphosphate sodium salt
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate

Specifications

White
208584-51-4
MFCD10567083
PHZDHEPQFXTBRQ-JBGYRJLXSA-N
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate
131875843
Lyophilized Powder
D-myo-Inositol-1,2-bisphosphate sodium salt
0.1mg
C6H14Na2O12P2
d-myo-inositol-1,2-diphosphate sodium salt
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na]
386.093
384.08
98%
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS