Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2,6-trisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,2,6-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD12912249
Synonym 1,2,6-IP3; Ins(1,2,6)-P3 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66721EXD

  SDS
alfa aesar™
J66721EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66721EXZ

  SDS
alfa aesar™
J66721EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-1,2,6-trisphosphate sodium salt
Physical Form Lyophilized Powder
Assay 98%
MDL Number MFCD12912249
Synonym 1,2,6-IP3; Ins(1,2,6)-P3 sodium salt
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C