Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt, 98%


Quantity 0.1mg

Manufacturer: alfa aesar™  J66647EXZ

 View more versions of this product

Catalog No. AAJ66647EXZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 588
Chemical Name or Material D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt
Assay 98%
Synonym 1,2,5,6-IP4 sodium salt; Ins(1,2,5,6)-P4 sodium salt
MDL Number MFCD12912294
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Quantity 0.1mg
Color White to off-white

Specifications

Chemical Name or Material D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt
Formula Weight 588
MDL Number MFCD12912294
Synonym 1,2,5,6-IP4 sodium salt; Ins(1,2,5,6)-P4 sodium salt
Accessories

-30° to -10°C

SDS & Safety Documents