Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt
Formula Weight 588
MDL Number MFCD12912294
Synonym 1,2,5,6-IP4 sodium salt; Ins(1,2,5,6)-P4 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66647EXZ

  SDS
alfa aesar™
J66647EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 588
Chemical Name or Material D-myo-Inositol-1,2,5,6-tetrakis(phosphate) sodium salt
Assay 98%
Synonym 1,2,5,6-IP4 sodium salt; Ins(1,2,5,6)-P4 sodium salt
MDL Number MFCD12912294
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Quantity 0.1mg
Color White to off-white
Safety and Handling

-30° to -10°C