Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2,4,5-tetrakis(phosphate) sodium salt

A useful IP3R activator

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,2,4,5-tetrakis(phosphate) sodium salt
Formula Weight 588
MDL Number MFCD12912299
Synonym 1,2,4,5-IP4 sodium salt; Ins(1,2,4,5)-P4 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66138EXZ

  SDS
alfa aesar™
J66138EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 588
Chemical Name or Material D-myo-Inositol-1,2,4,5-tetrakis(phosphate) sodium salt
Assay 98%
MDL Number MFCD12912299
Synonym 1,2,4,5-IP4 sodium salt; Ins(1,2,4,5)-P4 sodium salt
Solubility Information Soluble in water
Color White to off-white
Physical Form Lyophilized Powder
Quantity 0.1mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C