Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,2,3,4-tetrakis(phosphate) sodium salt, 98%


Quantity 0.1mg

Manufacturer: alfa aesar™  J66978EXZ

 View more versions of this product

Catalog No. AAJ66978EXZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 588
Chemical Name or Material D-myo-Inositol-1,2,3,4-tetrakis(phosphate) sodium salt
Physical Form Lyophilized Powder
Assay 98%
MDL Number MFCD12912296
Synonym 1,2,3,4-IP4 sodium salt; Ins(1,2,3,4)-P4 sodium salt
Quantity 0.1mg
Storage Note 1 -30° to -10°C
Color White to off-white

Specifications

Chemical Name or Material D-myo-Inositol-1,2,3,4-tetrakis(phosphate) sodium salt
Formula Weight 588
MDL Number MFCD12912296
Synonym 1,2,3,4-IP4 sodium salt; Ins(1,2,3,4)-P4 sodium salt
Accessories

-30° to -10°C