missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Methionine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6342206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-Methionine methyl ester hydrochloride | |
C6H14ClNO2S | |
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride | |
Cl.COC(=O)[C@H](N)CCSC | |
199.69 | |
199.7 | |
5 g |
69630-60-0 | |
MFCD00070385 | |
MEVUPUNLVKELNV-NUBCRITNSA-N | |
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride | |
12888616 | |
98% | |
Solid |
Chemical Identifiers
69630-60-0 | |
199.69 | |
MEVUPUNLVKELNV-NUBCRITNSA-N | |
12888616 | |
Cl.COC(=O)[C@H](N)CCSC |
C6H14ClNO2S | |
MFCD00070385 | |
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride | |
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only