missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Methionine ethyl ester hydrochloride, 95%

$87.76 - $1,107.24

Chemical Identifiers

CAS 7512-43-8
Molecular Formula C7H16ClNO2S
Molecular Weight (g/mol) 213.72
MDL Number MFCD00153451
InChI Key KPWCQEUBJAIORR-FYZOBXCZSA-N
Synonym h-d-met-oet.hcl, h-d-met-oet hcl, d-methionine ethyl ester hydrochloride, r-ethyl 2-amino-4-methylthio butanoate hydrochloride, ethyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, c7h15no2s.hcl, h-d-met-oethc, h-d-met-oet?cl, h-d-met-oet. hcl, d-methionine ethylester hydrochloride
PubChem CID 44717125
IUPAC Name ethyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES CCOC(=O)C(CCSC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6546106 View Documents Alfa Aesar™
H6546106
5g Each for $87.76
 
AAH6546114 View Documents Alfa Aesar™
H6546114
25g Each for $337.73
 
AAH6546122 View Documents Alfa Aesar™
H6546122
100g Each for $1,107.24
 
Specifications

Chemical Identifiers

7512-43-8
213.72
KPWCQEUBJAIORR-FYZOBXCZSA-N
44717125
CCOC(=O)C(CCSC)N.Cl
C7H16ClNO2S
MFCD00153451
h-d-met-oet.hcl, h-d-met-oet hcl, d-methionine ethyl ester hydrochloride, r-ethyl 2-amino-4-methylthio butanoate hydrochloride, ethyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, c7h15no2s.hcl, h-d-met-oethc, h-d-met-oet?cl, h-d-met-oet. hcl, d-methionine ethylester hydrochloride
ethyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride

Specifications

7512-43-8
MFCD00153451
KPWCQEUBJAIORR-FYZOBXCZSA-N
ethyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
44717125
Solid
5g
C7H16ClNO2S
h-d-met-oet.hcl, h-d-met-oet hcl, d-methionine ethyl ester hydrochloride, r-ethyl 2-amino-4-methylthio butanoate hydrochloride, ethyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, c7h15no2s.hcl, h-d-met-oethc, h-d-met-oet?cl, h-d-met-oet. hcl, d-methionine ethylester hydrochloride
CCOC(=O)C(CCSC)N.Cl
213.72
213.72
95%
D-Methionine ethyl ester hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C