Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Menthol, 99%

$62.39 - $185.34

Chemical Identifiers

Chemical Name or Material D-Menthol
CAS 15356-60-2
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00062983
Beilstein 1902292
Synonym (1S,2R,5S)-2-Isopropyl-5-methylcyclohexanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0610214

  SDS
alfa aesar™
L0610214
25g Each for $62.39

AAL0610222

  SDS
alfa aesar™
L0610222
100g Each for $185.34
Description & Specifications

Specifications

Formula Weight 156.27
Chemical Name or Material D-Menthol
Assay 99%
MDL Number MFCD00062983
Synonym (1S,2R,5S)-2-Isopropyl-5-methylcyclohexanol
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Molecular Formula C10H20O
CAS 15356-60-2
Beilstein 1902292
Health Hazard 1 Danger
RTECS Number OT0350000
Flash Point 91°C (195°F)
Optical Rotation +49° (c=10 in Ethanol)
Boiling Point 104° to 105°C (10mmHg)
Melting Point 41° to 44°C
Quantity 25g

Applications
D-Menthol is used for oral gel patch or film containing herb extracts or medicine, fruit extract, spearmint, and menthol for smoking cessation. It is used as flavoring in foods, cigarettes, liqueurs, cosmetics, and perfumes.

Solubility
Soluble in methanol (almost transparency), chloroform, alcohols, water (456 mg/L at 25°C), and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.