missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Melibiose Monohydrate, TCI America™

Manufacturer:  TCI America M005010G

 View more versions of this product

Catalog No. M005010G



Specifications

Specifications

D-(+)-Melibiose Monohydrate
185°C
66009-10-7
MFCD00198188
CHIDEFLSUMQFBY-CQIZOMOZSA-N
(2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
71308738
Crystal-Powder at 20°C
White
10g
C12H24O12
d +-melibiose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate
C(C1C(C(C(C(O1)OCC(C(C(C(C=O)O)O)O)O)O)O)O)O.O
360.312
342.30
≥99.0% (HPLC)

Chemical Identifiers

66009-10-7
360.312
CHIDEFLSUMQFBY-CQIZOMOZSA-N
71308738
C(C1C(C(C(C(O1)OCC(C(C(C(C=O)O)O)O)O)O)O)O)O.O
C12H24O12
MFCD00198188
d +-melibiose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate
(2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents