Please login to your online account to display your discounted pricing

D(+)-Melezitose monohydrate, ≥99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  225640250

 View more versions of this product

Catalog No. AC225640250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 522.45
Quantity 25g
Color White
Specific Rotation Condition +88° (20°C c=4,H2O)
TSCA TSCA
EINECS Number 209-894-9
Merck Index 14, 5819
Beilstein 31, 466
Specific Rotation +86° to +90° (20°C, 589nm) (c=4, water)
Heavy Metals (as Pb) 5ppm max.
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C18H32O16·H2O
MDL Number MFCD00149448
CAS 10030-67-8
Chemical Name or Material D(+)-Melezitose monohydrate

Specifications

Chemical Name or Material D(+)-Melezitose monohydrate
Name Note 99+%
CAS 10030-67-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C18H32O16·H2O
Formula Weight 522.45
MDL Number MFCD00149448