Please login to your online account to display your discounted pricing

D(+)-Melezitose monohydrate, ≥99%, ACROS Organics™

$34.64 - $134.57

Chemical Identifiers

Chemical Name or Material D(+)-Melezitose monohydrate
Name Note 99+%
CAS 10030-67-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C18H32O16·H2O
Formula Weight 522.45
MDL Number MFCD00149448
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC225640050

  SDS
thermo fisher scientific
225640050
Glass bottle 5g Each for $34.64

AC225640250

  SDS
fisher scientific acros
225640250
Glass bottle 25g Each for $134.57
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 522.45
Quantity 5g
Color White
Beilstein 31, 466
Merck Index 14, 5819
Specific Rotation Condition +88° (20°C c=4,H2O)
EINECS Number 209-894-9
TSCA TSCA
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +86° to +90° (20°C, 589nm) (c=4, water)
Chemical Name or Material D(+)-Melezitose monohydrate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C18H32O16·H2O
MDL Number MFCD00149448
CAS 10030-67-8