missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Melezitose Hydrate, TCI America™

Manufacturer:  TCI America M00495G

 View more versions of this product

Catalog No. M00495G



Specifications

Specifications

D-(+)-Melezitose Hydrate
153°C
207511-10-2
MFCD00149448
CFXAVQUXSYFPDE-HCCWNPJRSA-N
(2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate
126963632
Crystal-Powder at 20°C
White
5g
C18H34O17
d-+-melezitose hydrate
C(C1C(C(C(C(O1)OC2C(C(OC2(CO)OC3C(C(C(C(O3)CO)O)O)O)CO)O)O)O)O)O.O
522.453
504.44
≥99.0% (GC)

Chemical Identifiers

207511-10-2
522.453
CFXAVQUXSYFPDE-HCCWNPJRSA-N
126963632
C(C1C(C(C(C(O1)OC2C(C(OC2(CO)OC3C(C(C(C(O3)CO)O)O)O)CO)O)O)O)O)O.O
C18H34O17
MFCD00149448
d-+-melezitose hydrate
(2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents