missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Mannose 98.0+%, TCI America™

Manufacturer:  TCI America M0045100G

 View more versions of this product

Catalog No. M0045100G



Specifications

Specifications

D-(+)-Mannose
100g
C6H12O6
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers
Crystal-Powder at 20°C
White
3458-28-4
MFCD00064122
180.16
≥98.0% (HPLC)

Chemical Identifiers

3458-28-4
C6H12O6
MFCD00064122
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0685

TSCA : Yes

Documents