missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Mannono-1,4-lactone 97.0+%, TCI America™
Supplier: TCI America M09585G
Specifications
D-Mannono-1,4-lactone | |
153°C | |
C6H10O6 | |
5 g | |
C(C(C1C(C(C(=O)O1)O)O)O)O | |
178.14 | |
178.14 | |
Crystalline Powder |
26301-79-1 | |
White | |
MFCD00065020 | |
SXZYCXMUPBBULW-SQOUGZDYSA-N | |
(3S,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one | |
1774581 | |
≥97.0% (GC) |
Chemical Identifiers
26301-79-1 | |
178.14 | |
SXZYCXMUPBBULW-SQOUGZDYSA-N | |
(3S,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one |
C6H10O6 | |
MFCD00065020 | |
1774581 | |
C(C(C1C(C(C(=O)O1)O)O)O)O |
Safety and Handling
TSCA : No