missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Mannitol, ACS reagent, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 423920050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-Mannitol | |
0.01% max. | |
Authentic | |
C6H14O6 | |
5kg | |
15, 5810 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
hexane-1,2,3,4,5,6-hexol | |
6251 | |
182.17 | |
0.01% max |
69-65-8 | |
0.05% max. (5 g, 105°C, 2 hrs) | |
Plastic drum | |
MFCD00064287 | |
01, IV, 2841 | |
+23.3° to +24.3° (25°C, 589nm) (Reagent ACS-directives) | |
Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents | |
OCC(O)C(O)C(O)C(O)CO | |
182.17 | |
CHEBI:16899 | |
ACS Reagent | |
0.0008 meq/g max |
Safety and Handling
EINECSNumber : 200-711-8
RUO â Research Use Only