missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Mannitol, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423920010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-Mannitol | |
165.0°C to 170.0°C | |
0.01% max. | |
Authentic | |
C6H14O6 | |
1 kg | |
15, 5810 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
hexane-1,2,3,4,5,6-hexol | |
6251 | |
182.17 | |
0.01% max | |
Crystalline Powder |
69-65-8 | |
White | |
0.05% max. (5 g, 105°C, 2 hrs) | |
Plastic bottle | |
MFCD00064287 | |
01, IV, 2841 | |
+23.3° to +24.3° (25°C, 589nm) (Reagent ACS-directives) | |
Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents | |
OCC(O)C(O)C(O)C(O)CO | |
182.17 | |
CHEBI:16899 | |
ACS Reagent | |
0.0008 meq/g max |
Chemical Identifiers
69-65-8 | |
182.17 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
6251 | |
hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
MFCD00064287 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
CHEBI:16899 | |
OCC(O)C(O)C(O)C(O)CO |
Safety and Handling
EINECSNumber : 200-711-8
RUO – Research Use Only