missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Mannitol, For ACS analysis, 98.4%, MP Biomedicals™

Manufacturer:  Mp Biomedicals 0215254091

 View more versions of this product

Catalog No. ICN15254091



Specifications

Specifications

D-Mannitol
White
For ACS analysis
0.984
cordycepic acid, d-mannitol, manna sugar, mannazucker, mannidex, mannit, mannite, mannitol, osmitrol, osmofundin
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:16899
Powder
1.52g/mL
1kg
69-65-8
C6H14O6
FBPFZTCFMRRESA-KVTDHHQDSA-N
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.2

Chemical Identifiers

69-65-8
182.172
cordycepic acid, d-mannitol, manna sugar, mannazucker, mannidex, mannit, mannite, mannitol, osmitrol, osmofundin
CHEBI:16899
C(C(C(C(C(CO)O)O)O)O)O
C6H14O6
FBPFZTCFMRRESA-KVTDHHQDSA-N
6251
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Documents