Please login to your online account to display your discounted pricing

D-Mannitol, For ACS analysis, 98.4%, MP Biomedicals™


Quantity 500g

Manufacturer: mp biomedicals  0215254090

 View more versions of this product

Catalog No. ICN15254090

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 182.2
Physical Form Powder
Color White
Quantity 500g
Density 1.52g/mL
Synonym 1,2,3,4,5,6-Hexahexol
Name Note For ACS analysis
Molecular Formula C6H14O6
CAS 69-65-8
Chemical Name or Material D-Mannitol
Assay Percent Range 0.984

Specifications

Chemical Name or Material D-Mannitol
Name Note For ACS analysis
CAS 69-65-8
Physical Form Powder
Assay Percent Range 0.984
Molecular Formula C6H14O6
Formula Weight 182.2
Synonym 1,2,3,4,5,6-Hexahexol