Please login to your online account to display your discounted pricing

D-Mannitol, For ACS analysis, 98.4%, MP Biomedicals™

$115.62 - $185.30

Chemical Identifiers

Chemical Name or Material D-Mannitol
Name Note For ACS analysis
CAS 69-65-8
Physical Form Powder
Assay Percent Range 0.984
Molecular Formula C6H14O6
Formula Weight 182.2
Synonym 1,2,3,4,5,6-Hexahexol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15254090

 
mp biomedicals
0215254090
500g Each for $115.62

ICN15254091

 
mp biomedicals
0215254091
1Kg Each for $185.30
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 182.2
Color White
Quantity 500g
Density 1.52g/mL
Chemical Name or Material D-Mannitol
Name Note For ACS analysis
Assay Percent Range 0.984
Molecular Formula C6H14O6
CAS 69-65-8
Synonym 1,2,3,4,5,6-Hexahexol