missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Mannitol, 98+%, ACROS Organics™

$33.33 - $289.45

Chemical Identifiers

CAS 69-65-8
Assay Percent Range 98+%
Molecular Formula C6H14O6
Formula Weight 182.17
MDL Number MFCD00064287
Chemical Name or Material D-Mannitol
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC125340010 SDS Thermo Fisher Scientific
125340010
Plastic bottle 1kg Each for $77.00
AC125340025 SDS Fisher Scientific Acros
AC125340025
Plastic bottle 2.5kg Each for $159.22
AC125341000 SDS Fisher Scientific Acros
AC125341000
Plastic bottle 100g Each for $33.33
AC125345000 SDS Thermo Fisher Scientific
125345000
Plastic bottle 500g Each for $48.05
AC125340050 SDS Thermo Fisher Scientific
AC125340050
Plastic bucket 5kg Each for $289.45
Specifications Description & Specifications

Chemical Identifiers

69-65-8
C6H14O6
MFCD00064287
≥98.0%
98+%
182.17
D-Mannitol

Specifications

White
1kg
Crystalline Powder
C6H14O6
182.17
01, IV, 2841
0.5ppm max.
1ppm max.
+23° to +25° (20°C, 589nm) (PHEUR)
≥98.0%
165° to 170°C
69-65-8
98+%
Authentic
MFCD00064287
15, 5810
0.3% max. (105°C, 4 hrs)
Plastic bottle
D-Mannitol