missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Malic Acid, TCI America™

Manufacturer:  TCI America M00215G

 View more versions of this product

Catalog No. M00215G



Specifications

Specifications

D-(+)-Malic Acid
103°C
636-61-3
MFCD00004245
BJEPYKJPYRNKOW-UWTATZPHSA-N
(2R)-2-hydroxybutanedioic acid
92824
134.09
≥98.0% (GC,T)
White
5g
C4H6O5
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, r-malic acid, l +-malic acid, d-malate
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:30796
Crystal-Powder at 20°C

Chemical Identifiers

636-61-3
134.087
BJEPYKJPYRNKOW-UWTATZPHSA-N
92824
(2R)-2-hydroxybutanedioic acid
C4H6O5
MFCD00004245
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, r-malic acid, l +-malic acid, d-malate
CHEBI:30796
C(C(C(=O)O)O)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1442

RTECSNumber : ON7260000

TSCA : No

Documents