missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(-)-Lyxose, TCI America™

Manufacturer:  TCI America L007325G

 View more versions of this product

Catalog No. L007325G



Specifications

Specifications

D-(-)-Lyxose
25g
C5H10O5
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
C(C(C(C(C=O)O)O)O)O
150.13
CHEBI:16789
Crystal-Powder at 20°C
White-Yellow
1114-34-7
MFCD00064362
PYMYPHUHKUWMLA-UOWFLXDJSA-N
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
65550
150.13
≥98.0% (HPLC)

Chemical Identifiers

1114-34-7
150.13
PYMYPHUHKUWMLA-UOWFLXDJSA-N
65550
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00064362
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
CHEBI:16789
C(C(C(C(C=O)O)O)O)O
Safety and Handling

Safety and Handling

TSCA : Yes

Documents