missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals D(-)-Lyxose, 99+%, mixture of anomers
$106.83 - $466.84
Chemical Identifiers
CAS | 1114-34-7 |
---|---|
Molecular Formula | C5H10O5 |
Molecular Weight (g/mol) | 150.13 |
MDL Number | MFCD00064362 |
InChI Key | PYMYPHUHKUWMLA-UOWFLXDJSA-N |
Synonym | 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci |
PubChem CID | 65550 |
ChEBI | CHEBI:16789 |
IUPAC Name | (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal |
SMILES | C(C(C(C(C=O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC205230050
|
Thermo Scientific Chemicals
205230050 |
5 g | Glass bottle |
Each for $106.83
|
|
||||
AC205230250
|
Thermo Scientific Chemicals
205230250 |
25 g | Glass bottle |
Each for $466.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1114-34-7 | |
150.13 | |
PYMYPHUHKUWMLA-UOWFLXDJSA-N | |
65550 | |
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal |
C5H10O5 | |
MFCD00064362 | |
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci | |
CHEBI:16789 | |
C(C(C(C(C=O)O)O)O)O |
Specifications
1114-34-7 | |
White to Yellow | |
99% min. (ELSD) (HPLC) | |
C5H10O5 | |
5 g | |
15, 5702 | |
− 14.00 | |
Solubility in water: freely soluble. Other solubilities: soluble in ethanol | |
C(C(C(C(C=O)O)O)O)O | |
150.13 | |
CHEBI:16789 | |
99+% | |
D(-)-Lyxose, Mixture of anomers |
108.0°C to 112.0°C | |
Authentic | |
Glass bottle | |
MFCD00064362 | |
31, 56 | |
− 14.00 (20.00°C c=4,H2O,NH3) | |
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci | |
PYMYPHUHKUWMLA-UOWFLXDJSA-N | |
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal | |
65550 | |
150.13 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 214-212-8
RUO – Research Use Only