missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
D-Lysine Methyl Ester Dihydrochloride, TCI America™
Chemical Identifiers
CAS |
67396-08-1 |
Molecular Formula |
C7H18Cl2N2O2 |
Molecular Weight (g/mol) |
233.133 |
MDL Number |
MFCD00039067 |
InChI Key |
SXZCBVCQHOJXDR-QYCVXMPOSA-N |
Synonym |
H-D-Lys-OMe.2HCl |
PubChem CID |
44629856 |
IUPAC Name |
methyl (2R)-2,6-diaminohexanoate;dihydrochloride |
SMILES |
COC(=O)C(CCCCN)N.Cl.Cl |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
67396-08-1 |
|
233.133 |
|
SXZCBVCQHOJXDR-QYCVXMPOSA-N |
|
44629856 |
|
COC(=O)C(CCCCN)N.Cl.Cl |
|
C7H18Cl2N2O2 |
|
MFCD00039067 |
|
H-D-Lys-OMe.2HCl |
|
methyl (2R)-2,6-diaminohexanoate;dihydrochloride |
Specifications
|
White |
|
5g |
|
C7H18Cl2N2O2 |
|
H-D-Lys-OMe.2HCl |
|
COC(=O)C(CCCCN)N.Cl.Cl |
|
233.133 |
|
233.13 |
|
≥98.0% (N,T) |
|
205°C |
|
67396-08-1 |
|
MFCD00039067 |
|
SXZCBVCQHOJXDR-QYCVXMPOSA-N |
|
methyl (2R)-2,6-diaminohexanoate;dihydrochloride |
|
44629856 |
|
Crystal-Powder at 20°C |
|
D-Lysine Methyl Ester Dihydrochloride |
Safety and Handling