Please login to your online account to display your discounted pricing

D-Luciferin sodium salt monohydrate, 98+%, ACROS Organics™


Quantity 10mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC336780100

 View more versions of this product

Catalog No. AC336780100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 320.32
Quantity 10mg
Color Yellow
Chemical Name or Material D-Luciferin sodium salt monohydrate
Assay Percent Range 98+%
Molecular Formula C11H7N2NaO3S2·H2O
MDL Number MFCD11865368
CAS 103404-75-7
Synonym Firefly Luciferin monosodium salt monohydrate
Infrared Spectrum Authentic
Health Hazard 1 MOISTURE SENSITIVE
Packaging Glass bottle
Health Hazard 2 AIR SENSITIVE
Specific Extinction 17500 min. (385nm, 0.5M Na2CO3) min. (pH 11.5)
Water 7% max. (K.F.)

Specifications

Chemical Name or Material D-Luciferin sodium salt monohydrate
CAS 103404-75-7
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C11H7N2NaO3S2·H2O
Formula Weight 320.32
MDL Number MFCD11865368
Synonym Firefly Luciferin monosodium salt monohydrate
Accessories

MOISTURE SENSITIVE

AIR SENSITIVE

SDS & Safety Documents