missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Limonene, 96.9%, MP Biomedicals
Manufacturer: Mp Biomedicals 0215523490
Specifications
D-Limonene | |
500mL | |
C10H16 | |
XMGQYMWWDOXHJM-JTQLQIEISA-N | |
(4R)-1-methyl-4-prop-1-en-2-ylcyclohexene | |
440917 | |
136.26 | |
0.969 |
Colorless to Yellow | |
5989-27-5 | |
+-4r-limonene, +-carvene, +-dipentene, +-limonene, +-p-mentha-1,8-diene, 4r-limonene, citrene, d-+-limonene, d-limonene, r-+-limonene | |
CC1=CCC(CC1)C(=C)C | |
136.238 | |
CHEBI:15382 | |
Liquid |
Chemical Identifiers
5989-27-5 | |
136.238 | |
+-4r-limonene, +-carvene, +-dipentene, +-limonene, +-p-mentha-1,8-diene, 4r-limonene, citrene, d-+-limonene, d-limonene, r-+-limonene | |
CHEBI:15382 | |
CC1=CCC(CC1)C(=C)C |
C10H16 | |
XMGQYMWWDOXHJM-JTQLQIEISA-N | |
440917 | |
(4R)-1-methyl-4-prop-1-en-2-ylcyclohexene |