missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Leucylglycylglycine 98.0+%, TCI America™

Manufacturer:  TCI America L0031100MG

Catalog No. L0031100MG



Specifications

Specifications

D-Leucylglycylglycine
100mg
C10H19N3O4
H-D-Leu-Gly-Gly-OH
CC(C)CC(C(=O)NCC(=O)NCC(=O)O)N
245.279
245.28
≥98.0% (HPLC,T)
White
18625-22-4
MFCD00021740
VWHGTYCRDRBSFI-SSDOTTSWSA-N
2-[[2-[[(2R)-2-amino-4-methylpentanoyl]amino]acetyl]amino]acetic acid
87731
Crystal-Powder at 20°C

Chemical Identifiers

18625-22-4
245.279
VWHGTYCRDRBSFI-SSDOTTSWSA-N
87731
CC(C)CC(C(=O)NCC(=O)NCC(=O)O)N
C10H19N3O4
MFCD00021740
H-D-Leu-Gly-Gly-OH
2-[[2-[[(2R)-2-amino-4-methylpentanoyl]amino]acetyl]amino]acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents