missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Leucinol, 97%

Manufacturer:  Alfa Aesar™ L1324303

 View more versions of this product

Catalog No. AAL1324303



Specifications

Specifications

D-Leucinol
198°C to 200°C (690mmHg)
90°C (194°F)
C6H15NO
1719241
VPSSPAXIFBTOHY-ZCFIWIBFSA-N
(2R)-2-amino-4-methylpentan-1-ol
2724002
97%
53448-09-2
1g
−3.2° (c=9 in Ethanol)
MFCD00004734
d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #, d-leucinol
CC(C)CC(CO)N
117.192
117.19

Chemical Identifiers

53448-09-2
117.192
VPSSPAXIFBTOHY-ZCFIWIBFSA-N
2724002
CC(C)CC(CO)N
C6H15NO
MFCD00004734
d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #, d-leucinol
(2R)-2-amino-4-methylpentan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents