missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Leucine tert-Butyl Ester Hydrochloride, TCI America™

$53.47 - $158.89

Chemical Identifiers

CAS 13081-32-8
Molecular Formula C10H22ClNO2
Molecular Weight (g/mol) 223.741
MDL Number MFCD00083642
InChI Key RFUWRXIYTQGFGA-DDWIOCJRSA-N
Synonym h-d-leu-otbu.hcl, r-tert-butyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-otbu hcl, d-leucine tert-butyl ester hydrochloride, d-leucine t-butyl ester hydrochloride, tert-butyl 2r-2-amino-4-methylpentanoate hydrochloride, r-tert-butyl 2-amino-4-methylpentanoate hcl, h-d-leu-otbu. hcl, d-leucine tert.butyl ester hydrochloride, d-leucine-t-butylester hydrochloride
PubChem CID 44629930
IUPAC Name tert-butyl (2R)-2-amino-4-methylpentanoate;hydrochloride
SMILES CC(C)CC(C(=O)OC(C)(C)C)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L01961G View Documents TCI America
L01961G
1g Each for $53.47
 
L01965G View Documents TCI America
L01965G
5g Each for $158.89
 
Specifications

Chemical Identifiers

13081-32-8
223.741
RFUWRXIYTQGFGA-DDWIOCJRSA-N
44629930
CC(C)CC(C(=O)OC(C)(C)C)N.Cl
C10H22ClNO2
MFCD00083642
h-d-leu-otbu.hcl, r-tert-butyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-otbu hcl, d-leucine tert-butyl ester hydrochloride, d-leucine t-butyl ester hydrochloride, tert-butyl 2r-2-amino-4-methylpentanoate hydrochloride, r-tert-butyl 2-amino-4-methylpentanoate hcl, h-d-leu-otbu. hcl, d-leucine tert.butyl ester hydrochloride, d-leucine-t-butylester hydrochloride
tert-butyl (2R)-2-amino-4-methylpentanoate;hydrochloride

Specifications

White-Yellow
13081-32-8
MFCD00083642
RFUWRXIYTQGFGA-DDWIOCJRSA-N
tert-butyl (2R)-2-amino-4-methylpentanoate;hydrochloride
44629930
Crystal-Powder at 20°C
D-Leucine tert-Butyl Ester Hydrochloride
1g
C10H22ClNO2
h-d-leu-otbu.hcl, r-tert-butyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-otbu hcl, d-leucine tert-butyl ester hydrochloride, d-leucine t-butyl ester hydrochloride, tert-butyl 2r-2-amino-4-methylpentanoate hydrochloride, r-tert-butyl 2-amino-4-methylpentanoate hcl, h-d-leu-otbu. hcl, d-leucine tert.butyl ester hydrochloride, d-leucine-t-butylester hydrochloride
CC(C)CC(C(=O)OC(C)(C)C)N.Cl
223.741
223.74
≥98.0% (N,T)
Safety and Handling

Safety and Handling

TSCA : No