missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Leucine, TCI America™

Manufacturer:  TCI America L002725G

 View more versions of this product

Catalog No. L002725G



Specifications

Specifications

D-Leucine
25g
C6H13NO2
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CC(C)CC(C(=O)O)N
131.175
CHEBI:28225
Crystal-Powder at 20°C
White
328-38-1
MFCD00063088
ROHFNLRQFUQHCH-RXMQYKEDSA-N
(2R)-2-amino-4-methylpentanoic acid
439524
131.18
≥98.0% (T)

Chemical Identifiers

328-38-1
131.175
ROHFNLRQFUQHCH-RXMQYKEDSA-N
439524
(2R)-2-amino-4-methylpentanoic acid
C6H13NO2
MFCD00063088
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CHEBI:28225
CC(C)CC(C(=O)O)N
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1632

RTECSNumber : OH2840000

TSCA : Yes

Documents