missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Leucine Methyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America L01981G

 View more versions of this product

Catalog No. L01981G



Specifications

Specifications

D-Leucine Methyl Ester Hydrochloride
151°C
5845-53-4
MFCD00066115
DODCBMODXGJOKD-FYZOBXCZSA-N
methyl (2R)-2-amino-4-methylpentanoate;hydrochloride
11600782
Crystal-Powder at 20°C
White
1g
C7H16ClNO2
d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893
CC(C)CC(C(=O)OC)N.Cl
181.66
181.66
≥98.0% (N,T)

Chemical Identifiers

5845-53-4
181.66
DODCBMODXGJOKD-FYZOBXCZSA-N
11600782
CC(C)CC(C(=O)OC)N.Cl
C7H16ClNO2
MFCD00066115
d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893
methyl (2R)-2-amino-4-methylpentanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents