missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Leucine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6545606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
151°C | |
5845-53-4 | |
C7H16ClNO2 | |
3655228 | |
DODCBMODXGJOKD-HQFIOVGONA-N | |
methyl (2R)-2-amino-4-methylpentanoate hydrochloride | |
11600782 | |
98% | |
Hygroscopic |
D-Leucine methyl ester hydrochloride | |
White | |
MFCD00066115 | |
d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893 | |
Cl.COC(=O)[C@H](N)CC(C)C | |
181.66 | |
181.66 | |
5 g | |
Solid |
Chemical Identifiers
5845-53-4 | |
181.66 | |
DODCBMODXGJOKD-HQFIOVGONA-N | |
11600782 | |
Cl.COC(=O)[C@H](N)CC(C)C |
C7H16ClNO2 | |
MFCD00066115 | |
d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893 | |
methyl (2R)-2-amino-4-methylpentanoate hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only