Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Leucine ethyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material D-Leucine ethyl ester hydrochloride
CAS 73913-65-2
Formula Weight 195.69
MDL Number MFCD00190936
Synonym H-D-Leu-OEt.HCl; (R)-Ethyl 2-amino-4-methylpentanoate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6591603

 
alfa aesar™
H6591603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6591614

 
alfa aesar™
H6591614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6591606

 
alfa aesar™
H6591606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.69
Melting Point 121° to 127°C
Assay 98%
Synonym H-D-Leu-OEt.HCl; (R)-Ethyl 2-amino-4-methylpentanoate hydrochloride
Chemical Name or Material D-Leucine ethyl ester hydrochloride
Physical Form Powder
MDL Number MFCD00190936
CAS 73913-65-2
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Sensitivity Hygroscopic
Safety and Handling

2° to 8°C