missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Leucine, 99%

Manufacturer:  Alfa Aesar™ A1484222

 View more versions of this product

Catalog No. AAA1484222



Description

Description

Applications
D-Leucine is used in the formation of sterols. It is also used as a selectable marker.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

D-Leucine
C6H13NO2
1721721
ROHFNLRQFUQHCH-RXMQYKEDSA-N
(2R)-2-amino-4-methylpentanoic acid
439524
131.18
>300°C
−15° (c=5 in 5N HCl)
328-38-1
MFCD00063088
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CC(C)CC(C(=O)O)N
131.175
CHEBI:28225
99%
100g

Chemical Identifiers

328-38-1
131.175
ROHFNLRQFUQHCH-RXMQYKEDSA-N
439524
(2R)-2-amino-4-methylpentanoic acid
C6H13NO2
MFCD00063088
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CHEBI:28225
CC(C)CC(C(=O)O)N
Safety and Handling

Safety and Handling

RTECSNumber : OH2840000

TSCA : TSCA

SDS Documents