Please login to your online account to display your discounted pricing

D-Lactose Monohydrate (Powder/NF), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLL6212

 View more versions of this product

Catalog No. L6-212

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 360.32
Melting Point 219°C
Quantity 2.5kg
Density 1.525g/cm³
Purity Grade NF
Physical Form Solid
Molecular Formula C12H22O11 · H2O
Alpha Vector Lactose Monohydrate
Decomposition Information >219°C
Color White
Escherichia coli (E coli) Negative Negative/Positive
Identification Pass Test
ChemAlert Storage Symbol Gray
Residual Solvents Meets Requirements
Total Microbial Count 100 CFU/g max.
Total Mold and Yeast 50 CFU/g max.
Water 4.5 to 5.5%
Clarity and Color Pass Test
Heavy Metals (as Pb) 5ppm max.
Ignition Residue 0.1% max.
MDL Number MFCD00150747
Synonym Milk Sugar
Loss on Drying 0.5% max.
Packaging Poly Bottle
Acidity Alkalinity Pass Test
Protein and Light Absorbing Substances Pass Test
Additional Information Vapor Pressure: Negligible
Specific Rotation 54.4 to 55.9° (+ or -)
Chemical Name or Material D-Lactose Monohydrate
CAS 5989-81-1

Specifications

Chemical Name or Material D-Lactose Monohydrate
CAS 5989-81-1
Purity Grade NF
Physical Form Solid
Molecular Formula C12H22O11 · H2O
Formula Weight 360.32
MDL Number MFCD00150747
Synonym Milk Sugar
Accessories