missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Lactose Monohydrate (Powder/NF/EP/BP/JP), Fisher Chemical

Manufacturer:  Thermo Scientific L812

 View more versions of this product

Catalog No. L8-12



Specifications

Specifications

0.04 AU max. (BP/EP) at 400nm, 0.04 AU max. at 400nm, 0.07 AU max. (BP/EP), 0.07 AU max. from 270 to 300nm, 0.25 AU max. (BP/EP), 0.25 AU max. from 210 to 220nm
Lactose Monohydrate
D-Lactose Monohydrate
1.525g/cm³
0.1% max. (JP/NF)
Poly Pail
54.4 to 55.9° (BP/EP/JP/NF)
Negligible
White
12kg
C12H24O12
d-lactose monohydrate, d-+-lactosemonohydrate
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
360.312
360.32
BP/EP/JP/NF
Pass Test (BP/EP/JP/NF)
Pass Test (BP/EP/JP/NF)
>219°C
Pass Test (EP/BP/NF)
0.5% max. (JP/NF)
Meets Requirements (BP/EP)
0.1% max. (BP/EP)
4.5 to 5.5% (BP/EP/JP/NF)
219°C
64044-51-5
MFCD00150747
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
133126686
Solid

Chemical Identifiers

64044-51-5
360.312
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
133126686
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
C12H24O12
MFCD00150747
d-lactose monohydrate, d-+-lactosemonohydrate
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Safety and Handling

Safety and Handling

SDS Documents