missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Lactose

$106.26 - $189.62

Chemical Identifiers

CAS 63-42-3
Molecular Formula C12H22O11
MDL Number MFCD00151251
Synonym Gal-beta-1,4-Glc; 4-O-beta-D-Galactopyranosyl-D-glucopyranose
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6637636 View Documents Alfa Aesar™
J6637636
500g Each for $106.26
 
AAJ66376A1 View Documents Alfa Aesar™
J66376A1
1kg Each for $189.62
 
Description

Description

Applications
D-Lactose is used for the culture of lactic acid metabolizing bacteria, lactic acid bacteria. D-Lactose is used to identify and characterized galectins.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Chemical Identifiers

63-42-3
C12H22O11
MFCD00151251
Gal-beta-1,4-Glc; 4-O-beta-D-Galactopyranosyl-D-glucopyranose

Specifications

Odorless
500g
C12H22O11
93796
Gal-beta-1,4-Glc; 4-O-beta-D-Galactopyranosyl-D-glucopyranose
Powder
White
63-42-3
MFCD00151251
14,5343
342.3
D-Lactose
Safety and Handling

Safety and Handling

RTECSNumber : OD9625000

TSCA : TSCA