Please login to your online account to display your discounted pricing

D(+)-Lactic acid, lithium salt, 98%, pure, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  215031000

 View more versions of this product

Catalog No. AC215031000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Purity Grade Extra Pure
Physical Form Fine Crystalline Powder
Formula Weight 96.01
Quantity 100mg
Color White
Beilstein 03, 267
Specific Rotation Condition +14.50° (20°C c=1,H2O)
EINECS Number 248-693-0
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material D(+)-Lactic acid, lithium salt
Name Note 98%
Assay Percent Range 98%
Molecular Formula C3H5O3Li
Linear Formula CH3CH(OH)CO2Li
MDL Number MFCD00013075
CAS 27848-81-3
Synonym Lithium D(+)lactate
Packaging Glass bottle
Specific Rotation +13° to +16° (20°C, 589nm) (c=1, H2O)

Specifications

Chemical Name or Material D(+)-Lactic acid, lithium salt
Name Note 98%
CAS 27848-81-3
Purity Grade Extra Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H5O3Li
Linear Formula CH3CH(OH)CO2Li
Formula Weight 96.01
MDL Number MFCD00013075
Synonym Lithium D(+)lactate
Accessories

HYGROSCOPIC

SDS & Safety Documents