Please login to your online account to display your discounted pricing

D(+)-Lactic acid, lithium salt, 98%, pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D(+)-Lactic acid, lithium salt
Name Note 98%
CAS 27848-81-3
Purity Grade Extra Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H5O3Li
Linear Formula CH3CH(OH)CO2Li
Formula Weight 96.01
MDL Number MFCD00013075
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215031000

  SDS
fisher scientific acros
215031000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 96.01
Purity Grade Extra Pure
Physical Form Fine Crystalline Powder
Quantity 100mg
Color White
Beilstein 03, 267
Specific Rotation Condition +14.50° (20°C c=1,H2O)
EINECS Number 248-693-0
Infrared Spectrum Authentic
Health Hazard 1 HYGROSCOPIC
Packaging Glass bottle
Specific Rotation +13° to +16° (20°C, 589nm) (c=1, H2O)
Synonym Lithium D(+)lactate
Molecular Formula C3H5O3Li
MDL Number MFCD00013075
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material D(+)-Lactic acid, lithium salt
Linear Formula CH3CH(OH)CO2Li
CAS 27848-81-3
Safety and Handling

HYGROSCOPIC