missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Homoserine, TCI America™

$171.11 - $571.41

Chemical Identifiers

CAS 6027-21-0
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00077786
InChI Key UKAUYVFTDYCKQA-GSVOUGTGSA-N
Synonym d-homoserine, 2r-2-amino-4-hydroxybutanoic acid, r-2-amino-4-hydroxybutanoic acid, r-+-2-amino-4-hydroxybutyric acid, h-d-hoser-oh, homoserine, d-homoserin, homoserine, d, ambotzhaa1028, pubchem13826
PubChem CID 2724170
ChEBI CHEBI:30654
IUPAC Name (2R)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12245G View Documents TCI America
H12245G
5g Each for $571.41
 
H12241G View Documents TCI America
H12241G
1g Each for $171.11
 
Specifications

Chemical Identifiers

6027-21-0
119.12
UKAUYVFTDYCKQA-GSVOUGTGSA-N
2724170
(2R)-2-amino-4-hydroxybutanoic acid
C4H9NO3
MFCD00077786
d-homoserine, 2r-2-amino-4-hydroxybutanoic acid, r-2-amino-4-hydroxybutanoic acid, r-+-2-amino-4-hydroxybutyric acid, h-d-hoser-oh, homoserine, d-homoserin, homoserine, d, ambotzhaa1028, pubchem13826
CHEBI:30654
C(CO)C(C(=O)O)N

Specifications

White-Yellow
5g
C4H9NO3
d-homoserine, 2r-2-amino-4-hydroxybutanoic acid, r-2-amino-4-hydroxybutanoic acid, r-+-2-amino-4-hydroxybutyric acid, h-d-hoser-oh, homoserine, d-homoserin, homoserine, d, ambotzhaa1028, pubchem13826
C(CO)C(C(=O)O)N
119.12
CHEBI:30654
Crystal-Powder at 20°C
D-Homoserine
204°C
6027-21-0
MFCD00077786
UKAUYVFTDYCKQA-GSVOUGTGSA-N
(2R)-2-amino-4-hydroxybutanoic acid
2724170
119.12
≥96.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2759

TSCA : No