Please login to your online account to display your discounted pricing

D(-)-Glutamine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D(-)-Glutamine
Name Note 98%
CAS 5959-95-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10N2O3
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Formula Weight 146.15
MDL Number MFCD00065607
Synonym D(-)-Glutamic acid-5-amide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC135160010

  SDS
fisher scientific acros
135160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC135160050

  SDS
fisher scientific acros
135160050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 146.15
Quantity 1g
Color White
Specific Rotation Condition −32.00 (20.00°C 589nm,c=10,N HCl)
Specific Rotation -30° to -34° (20°C, 589nm) (c=10, 2N HCl)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym D(-)-Glutamic acid-5-amide
MDL Number MFCD00065607
Molecular Formula C5H10N2O3
Name Note 98%
Linear Formula H2NCOCH2CH2CH(NH2)COOH
CAS 5959-95-5
Chemical Name or Material D(-)-Glutamine
Assay Percent Range 98%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.