missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6346303
Description
It is employed as a intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
D-Glutamic acid 5-tert-butyl ester | |
C9H17NO4 | |
h-d-glu otbu-oh, r-2-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamicacid, 5-1,1-dimethylethyl ester, 2r-2-amino-5-tert-butoxy-5-oxopentanoic acid, ambotzhaa1231, l-glutamicacid5-tert-butylester, r-2-amino-5-tert-butoxy-5-oxopentanoicacid | |
OIOAKXPMBIZAHL-ZCFIWIBFSA-N | |
(2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid | |
7016383 | |
95% | |
Solid |
45125-00-6 | |
MFCD00153437 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)CCC(C(=O)O)N | |
203.238 | |
203.24 | |
1 g |
Chemical Identifiers
45125-00-6 | |
203.238 | |
OIOAKXPMBIZAHL-ZCFIWIBFSA-N | |
7016383 | |
CC(C)(C)OC(=O)CCC(C(=O)O)N |
C9H17NO4 | |
MFCD00153437 | |
h-d-glu otbu-oh, r-2-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamicacid, 5-1,1-dimethylethyl ester, 2r-2-amino-5-tert-butoxy-5-oxopentanoic acid, ambotzhaa1231, l-glutamicacid5-tert-butylester, r-2-amino-5-tert-butoxy-5-oxopentanoicacid | |
(2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only