missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Glutamic acid 1-tert-butyl ester, 98%

Manufacturer:  Alfa Aesar™ H63318MD

 View more versions of this product

Catalog No. AAH63318MD



Description

Description

Applications
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

D-Glutamic acid 1-tert-butyl ester
C9H17NO4
h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
CC(C)(C)OC(=O)C(CCC(=O)O)N
203.238
203.24
98%
250mg
25456-76-2
MFCD09842819
QVAQMUAKTNUNLN-ZCFIWIBFSA-N
(4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
7019726
Powder
White

Chemical Identifiers

25456-76-2
203.238
QVAQMUAKTNUNLN-ZCFIWIBFSA-N
7019726
CC(C)(C)OC(=O)C(CCC(=O)O)N
C9H17NO4
MFCD09842819
h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
(4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents