missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Glucurono-6,3-lactone acetonide, 98+%

$82.96 - $282.20

Chemical Identifiers

CAS 20513-98-8
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00061641
InChI Key BDBGJSXZKMTMGP-BOFVIFSBSA-N
Synonym d-glucurono-6,3-lactone acetonide
PubChem CID 134129430
IUPAC Name (3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one
SMILES CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1776509 View Documents Alfa Aesar™
L1776509
10g Each for $82.96
 
AAL1776518 View Documents Alfa Aesar™
L1776518
50g Each for $282.20
 
Specifications

Chemical Identifiers

20513-98-8
216.189
BDBGJSXZKMTMGP-BOFVIFSBSA-N
134129430
CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C
C9H12O6
MFCD00061641
d-glucurono-6,3-lactone acetonide
(3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one

Specifications

+52° (c=1 in Chloroform)
10g
C9H12O6
d-glucurono-6,3-lactone acetonide
CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C
216.189
216.19
D-Glucurono-6,3-lactone acetonide
119°C to 121°C
20513-98-8
MFCD00061641
BDBGJSXZKMTMGP-BOFVIFSBSA-N
(3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one
134129430
≥98%