missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Glucurono-3,6-lactone, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 225920250

 View more versions of this product

Catalog No. AC225920250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(10% aq. soln.) clear, colorless
Passes Tests
0.5% max. (80°C)
+19.00 (20.00°C c=10,H2O)
0.1% max.
170.0°C to 176.0°C
99+%
99+%
MFCD00135622
d-glucuro-3,6-lactone
UYUXSRADSPPKRZ-KKQCNMDGSA-N
(2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
91634076
Crystals or Crystalline Powder
D(+)-Glucurono-3, 6-lactone
Authentic
Plastic bottle
+ 19.00
White
25g
32449-92-6
C6H8O6
15, 4503
(5% in water) Clear colorless to light yellow
C(=O)C(C1C(C(C(=O)O1)O)O)O
176.12
176.12
99+%

Chemical Identifiers

32449-92-6
176.12
UYUXSRADSPPKRZ-KKQCNMDGSA-N
91634076
C(=O)C(C1C(C(C(=O)O1)O)O)O
C6H8O6
MFCD00135622
d-glucuro-3,6-lactone
(2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
Safety and Handling

Safety and Handling

EINECSNumber : 251-053-3

RTECSNumber : LZ8930000

TSCA : TSCA

SDS Documents